Learn More
Trajan Scientific Syringe Metal 10uL Removable Needle 6/PK
SDP
Catalog No. 5000494943
Encompass_Preferred
10 L removable needle syringe with flexible plunger and 5 cm 0 47 mm OD bevel tipped needle PK6
Catalog No. 50-004-94943 Supplier Trajan Scientific Supplier No. 002180
10 L removable needle syringe with flexible plunger and 5 cm 0 47 mm OD bevel tipped needle PK6