Learn More
Trajan Scientific Syringe Metal 250uL Removable Needle 1/EA
SDP
Catalog No. 5000494601
Encompass_Preferred
250 L removable needle syringe with 5 1 cm 0 72 mm OD LC tipped needle
Catalog No. 50-004-94601 Supplier Trajan Scientific Supplier No. 006310
250 L removable needle syringe with 5 1 cm 0 72 mm OD LC tipped needle