Learn More
Trajan Scientific Syringe Metal 250uL Removable Needle 1/EA
SDP
Catalog No. 5000494850
Encompass_Preferred
250 L removable needle syringe with 5 cm 0 5 mm OD bevel tipped needle
Catalog No. 50-004-94850 Supplier Trajan Scientific Supplier No. 006050
Add to Cart
Add to Cart
250 L removable needle syringe with 5 cm 0 5 mm OD bevel tipped needle