Learn More
Trajan Scientific Syringe Metal 25uL Fixed Needle 1/EA
SDP
Catalog No. 5000494787
Encompass_Preferred
25 L fixed needle syringe with 5 1 cm 0 72 mm OD LC tipped needle
Catalog No. 50-004-94787 Supplier Trajan Scientific Supplier No. 003300
25 L fixed needle syringe with 5 1 cm 0 72 mm OD LC tipped needle