Learn More
Trajan Scientific Syringe Metal 500uL Fixed Needle 1/EA
SDP
Catalog No. 5000494760
Encompass_Preferred
500 L fixed needle syringe with and 5 1 cm 0 72 mm OD LC tipped needle
Catalog No. 50-004-94760 Supplier Trajan Scientific Supplier No. 007300
500 L fixed needle syringe with and 5 1 cm 0 72 mm OD LC tipped needle