Learn More
Trajan Scientific Syringe Metal 500uL Removable Needle 1/EA
SDP
Catalog No. 5000494727
Encompass_Preferred
500 L removable needle syringe with 5 cm 0 5 mm OD bevel tipped needle
Catalog No. 50-004-94727 Supplier Trajan Scientific Supplier No. 007050
500 L removable needle syringe with 5 cm 0 5 mm OD bevel tipped needle