Learn More
Trajan Scientific Syringe Metal 50uL Fixed Needle 1/EA
SDP
Catalog No. 5000494387
Encompass_Preferred
50 L fixed needle syringe with 5 1 cm 0 72 mm OD LC tipped needle
Catalog No. 50-004-94387 Supplier Trajan Scientific Supplier No. 004300
50 L fixed needle syringe with 5 1 cm 0 72 mm OD LC tipped needle