Learn More
Trajan Scientific Syringe Metal 5uL Fixed Needle 1/EA
SDP
Supplier:  Trajan Scientific 001800
Encompass_Preferred
5 L fixed needle Agilent syringe with 4 2 cm 0 47 mm OD cone tipped needle
Catalog No. 50-004-93948