Learn More
Trajan Scientific Syringe Metal 5uL Fixed Needle 1/EA
SDP
Catalog No. 5000494838
Encompass_Preferred
5 L fixed needle CTC Analytics and Thermo Scientific syringe with 5 cm 0 63 mm OD cone tipped needle
Catalog No. 50-004-94838 Supplier Trajan Scientific Supplier No. 001981
5 L fixed needle CTC Analytics and Thermo Scientific syringe with 5 cm 0 63 mm OD cone tipped needle