Learn More
Trajan Scientific Syringe Metal 5uL Fixed Needle 1/EA
SDP
Supplier:  Trajan Scientific 001861
Encompass_Preferred
5 L fixed needle CTC Analytics PAL RTC (PAL3) Shimadzu AOC-6000 Thermo Scientific TriPlus RSH and Agilent PAL3 syringe with 5 7 cm 0 63 mm OD cone tipped needle
Catalog No. 50-004-94870