Learn More
Trajan Scientific Syringe Metal 5uL Fixed Needle 1/EA
SDP
Supplier:  Trajan Scientific 0019811
Encompass_Preferred
5 L fixed needle CTC Analytics Thermo Scientific AS3000 AS2000 SCION CP-8410 and CP-8400 syringe with 5 cm 0 63 mm OD cone tipped needle
Catalog No. 50-004-94262