Learn More
Trajan Scientific Syringe Metal 5uL Fixed Needle 1/EA
SDP
Catalog No. 5000494413
Encompass_Preferred
5 L fixed needle syringe with 5 cm 0 47 mm OD bevel tipped needle
Catalog No. 50-004-94413 Supplier Trajan Scientific Supplier No. 001000
5 L fixed needle syringe with 5 cm 0 47 mm OD bevel tipped needle