Learn More
Trajan Scientific Syringe Metal 5uL Removable Needle 1/EA
SDP
Supplier:  Trajan Scientific 001877
Encompass_Preferred
5 L removable needle CTC RTC and Thermo RSH autosampler syringe with 8 5 cm 26 gauge 0 47 mm OD cone tipped needle
Catalog No. 50-004-94758