Learn More
Trajan Scientific Syringe Metal 5uL Removable Needle 1/EA
SDP
Catalog No. 5000494812
Encompass_Preferred
5 L removable needle CTC Analytics PAL RTC (PAL3) Shimadzu AOC-6000 Thermo Scientific TriPlus RSH and Agilent PAL3 syringe with 5 7 cm 0 63 mm OD cone tipped needle
Catalog No. 50-004-94812 Supplier Trajan Scientific Supplier No. 001875
5 L removable needle CTC Analytics PAL RTC (PAL3) Shimadzu AOC-6000 Thermo Scientific TriPlus RSH and Agilent PAL3 syringe with 5 7 cm 0 63 mm OD cone tipped needle