Learn More
Trajan Scientific Syringe Metal 5uL Removable Needle 1/EA
SDP
Catalog No. 5000493960
Encompass_Preferred
5 L removable needle CTC Analytics and Thermo Scientific syringe with 5 cm 0 63 mm OD cone tipped needle
Catalog No. 50-004-93960 Supplier Trajan Scientific Supplier No. 001984
5 L removable needle CTC Analytics and Thermo Scientific syringe with 5 cm 0 63 mm OD cone tipped needle