Learn More
Trajan Scientific Syringe Metal 5uL Removable Needle 1/EA
SDP
Supplier:  Trajan Scientific 001495
Encompass_Preferred
5 L removable needle guided plunger syringe with 5 cm 0 47 mm OD bevel tipped needle (half scale)
Catalog No. 50-004-94944