Learn More
Trajan Scientific Syringe Nanovolume 0.5uL B style 1/EA
SDP
Supplier:  Trajan Scientific 000492
Encompass_Preferred
0 5 L NanoVolume CTC Analytics and Thermo Scientific syringe with 5 cm 0 63 mm OD cone tipped needle
Catalog No. 50-004-94377