Learn More
Trajan Scientific Syringe Nanovolume 0.5uL B style 1/EA
SDP
Catalog No. 5000494759
Encompass_Preferred
0 5 L NanoVolume syringe with 5 cm 0 47 mm OD cone tipped needle
Catalog No. 50-004-94759 Supplier Trajan Scientific Supplier No. 000303
0 5 L NanoVolume syringe with 5 cm 0 47 mm OD cone tipped needle