Learn More
Trajan Scientific Syringe Nanovolume 0.5uL B style 1/EA
SDP
Catalog No. 5000494840
Encompass_Preferred
0 5 L NanoVolume on-column syringe with 7 cm 0 47 mm OD needle
Catalog No. 50-004-94840 Supplier Trajan Scientific Supplier No. 000376
0 5 L NanoVolume on-column syringe with 7 cm 0 47 mm OD needle