Learn More
Trajan Scientific Syringe Nanovolume 0.5uL B style 1/EA
SDP
Catalog No. 5000493944
Encompass_Preferred
0 5 L NanoVolume syringe with 5 cm 0 47 mm OD cone tipped needle and repeating adaptor
Catalog No. 50-004-93944 Supplier Trajan Scientific Supplier No. 000353
0 5 L NanoVolume syringe with 5 cm 0 47 mm OD cone tipped needle and repeating adaptor