Learn More
Trajan Scientific Syringe Nanovolume 0.5uL B style 1/EA
SDP
Catalog No. 5000493946
Encompass_Preferred
0 5 L NanoVolume syringe with 7 cm 0 63 mm OD cone tipped needle and repeating adaptor
Catalog No. 50-004-93946 Supplier Trajan Scientific Supplier No. 000355
0 5 L NanoVolume syringe with 7 cm 0 63 mm OD cone tipped needle and repeating adaptor