Learn More
Trajan Scientific Syringe Nanovolume 0.5uL B style 1/EA
SDP
Catalog No. 5000494747
Encompass_Preferred
0 5 L NanoVolume on-column syringe with 7 5 cm 0 23 mm OD needle
Catalog No. 50-004-94747 Supplier Trajan Scientific Supplier No. 000380
0 5 L NanoVolume on-column syringe with 7 5 cm 0 23 mm OD needle