Learn More
Trajan Scientific Syringe Nanovolume 1uL B style 1/EA
SDP
Supplier:  Trajan Scientific 000553
Encompass_Preferred
1 0 L NanoVolume syringe with 7 cm 0 63 mm OD cone tipped needle and repeating adaptor
Catalog No. 50-004-94796