Learn More
Trajan Scientific Syringe Nanovolume 5uL B style 1/EA
SDP
Catalog No. 5000494511
Encompass_Preferred
5 0 L NanoVolume syringe with 7 cm 0 63 mm OD bevel tipped needle
Catalog No. 50-004-94511 Supplier Trajan Scientific Supplier No. 000803
5 0 L NanoVolume syringe with 7 cm 0 63 mm OD bevel tipped needle