Learn More
Trajan Scientific Syringe Nanovolume 5uL B style 1/EA
SDP
Catalog No. 5000494676
Encompass_Preferred
5 0 L NanoVolume syringe with 5 cm 0 63 mm OD bevel tipped needle
Catalog No. 50-004-94676 Supplier Trajan Scientific Supplier No. 000801
5 0 L NanoVolume syringe with 5 cm 0 63 mm OD bevel tipped needle