Learn More
Trajan Scientific Syringe Nanovolume 5uL B style 1/EA
SDP
Catalog No. 5000494591
Encompass_Preferred
5 0 L NanoVolume syringe with 5 cm 0 63 mm OD cone tipped needle
Catalog No. 50-004-94591 Supplier Trajan Scientific Supplier No. 000800
5 0 L NanoVolume syringe with 5 cm 0 63 mm OD cone tipped needle