Learn More
Trajan Scientific Syringe Nanovolume Needle Kit Repair Kit 1/EA
SDP
Catalog No. 5000494488
Encompass_Preferred
Replacemenet needle and plunger for 1 L NanoVolume syringe with 70 mm cone-tipped needle
Catalog No. 50-004-94488 Supplier Trajan Scientific Supplier No. 034057
Replacemenet needle and plunger for 1 L NanoVolume syringe with 70 mm cone-tipped needle