Learn More
Trajan Scientific Syringe Nanovolume Needle Kit Repair Kit 1/EA
SDP
Catalog No. 5000494427
Encompass_Preferred
Replacemenet needle and plunger for 0 5 L NanoVolume syringe with 7 5 cm 0 23 mm OD on-column needle
Catalog No. 50-004-94427 Supplier Trajan Scientific Supplier No. 033620
Add to Cart
Add to Cart
Replacemenet needle and plunger for 0 5 L NanoVolume syringe with 7 5 cm 0 23 mm OD on-column needle