Learn More
Trajan Scientific Syringe Nanovolume Needle Kit Repair Kit 1/EA
SDP
Catalog No. 5000494444
Encompass_Preferred
Replacemenet needle and plunger for 1 L Agilent auto-sampler NanoVolume syringe with 23-gauge needle
Catalog No. 50-004-94444 Supplier Trajan Scientific Supplier No. 034715
Replacemenet needle and plunger for 1 L Agilent auto-sampler NanoVolume syringe with 23-gauge needle