Learn More
Trajan Scientific Syringe Nanovolume Needle Kit Repair Kit 1/EA
SDP
Catalog No. 5000494824
Encompass_Preferred
Replacemenet needle and plunger for 0 5 L NanoVolume syringe with 50 mm 0 63 mm OD cone-tipped needle
Catalog No. 50-004-94824 Supplier Trajan Scientific Supplier No. 033010
Replacemenet needle and plunger for 0 5 L NanoVolume syringe with 50 mm 0 63 mm OD cone-tipped needle