Learn More
Trajan Scientific Syringe Nanovolume Needle Kit Repair Kit 1/EA
SDP
Catalog No. 5000494287
Encompass_Preferred
Replacemenet needle and plunger for 1 L NanoVolume syringe with 70 mm cone-tipped 26-gauge needle
Catalog No. 50-004-94287 Supplier Trajan Scientific Supplier No. 034610
Replacemenet needle and plunger for 1 L NanoVolume syringe with 70 mm cone-tipped 26-gauge needle