Learn More
Andwin Scientific SYRINGE NEEDLE 20 GAUGE PTFE
SDP
Catalog No. NC9170911
Encompass
The PTFE syringe tubing has a gauge of 20 and a measured length of 24 inches. The PTFE needles must be punctured with a hollow SS needle. Afterwards, the PTFE needle is then threaded through. The SS needle is withdrawn, permitting the septum to seal around the PTFE needle.
Catalog No. NC9170911 Supplier Andwin Scientific Supplier No. Z117374
The PTFE syringe tubing has a gauge of 20 and a measured length of 24 inches. The PTFE needles must be punctured with a hollow SS needle. Afterwards, the PTFE needle is then threaded through. The SS needle is withdrawn, permitting the septum to seal around the PTFE needle.