Learn More
Med Vet International Dynarex Syringe With Needle - Luer Lock, 21g, 1" Needle, 3mL, 800/Case
Supplier: Med Vet International 6997
Dynarex Syringes with Needles are designed to help deliver medication doses efficiently and accurately. The Syringes are equipped with double-beveled, thin-walled needles for smooth insertion and improved patient comfort. The Syringes feature smooth-gliding plungers with integrated stops to help prevent accidental needle pop-out. Each Syringe features a transparent barrel with bold, easy-to-read markings to allow for easy visualization and accurate measurement. Dynarex Syringes with Needles are compatible with luer lock and luer slip connections, and are color-coded for easy identification of needle gauge. Dynarex Syringes with Needles are designed to help provide accurate and effective medication dosing. Each disposable Syringe includes a sterile, stainless-steel needle with a double-beveled tip for improved patient comfort. The Syringes feature transparent barrels with bold, easy-to-read numbering and either luer lock or luer slip connections.