Learn More
Med Vet International 10Cc Nylon Syringe
Catalog No. 502832294
Encompass_Preferred
Virtually unbreakable nylon with transparent barrel, economically priced with easy-to-read black etched dose markings. Combination of luer lock and luer slip adaptor. NY, CT, IL, NJ, NH, DE and MN restrict the sale of syringes and needles to licensed professionals and entities. By proceeding to purchase through our website, you acknowledge that you are over 18 years of age.
Catalog No. 50-283-2294 Supplier Med Vet International Supplier No. JOR111AN
Virtually unbreakable nylon with transparent barrel, economically priced with easy-to-read black etched dose markings. Combination of luer lock and luer slip adaptor. NY, CT, IL, NJ, NH, DE and MN restrict the sale of syringes and needles to licensed professionals and entities. By proceeding to purchase through our website, you acknowledge that you are over 18 years of age.