Learn More
WELCH MATERIALS INC Welchrom Syringe Filter, NYLON, 25mm, 0.22μm, Prefilter, Double layers,100pcs/pk,10pk/cs.
Catalog No. NC2052762
Encompass_Preferred
Syringe Filter, NYLON, 25mm, 0.22μm, Prefilter, Double layers,100pcs/pk,10pk/cs.
Catalog No. NC2052762 Supplier WELCH MATERIALS INC Supplier No. S25NY022B10
Syringe Filter, NYLON, 25mm, 0.22μm, Prefilter, Double layers,100pcs/pk,10pk/cs.