Learn More
Med Vet International Becton Dickinson Syringe, 10mL, Luer Slip, 100/Box, 305219
Catalog No. 502831594
Encompass_Preferred
BD 305219 10mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 10mL Syringe fits conventional and safety needles. This quantity is 100/BX
Catalog No. 50-283-1594 Supplier Med Vet International Supplier No. BD305219Z
BD 305219 10mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 10mL Syringe fits conventional and safety needles. This quantity is 100/BX