Learn More
Med Vet International Standard Oral Syringe, 10mL Or 2Tsp, Clear, 100/Box
Catalog No. 502829899
Encompass_Preferred
Polypropylene syringes with separate ribbed tip caps. Dual graduations in milliliters and teaspoons. Oral feeding Luer tip (will not accept hypodermic needle)
Catalog No. 50-282-9899 Supplier Med Vet International Supplier No. SWD907102Z
Polypropylene syringes with separate ribbed tip caps. Dual graduations in milliliters and teaspoons. Oral feeding Luer tip (will not accept hypodermic needle)