Learn More
WELCH MATERIALS INC Welchrom Syringe Filter, PES, 13mm, 0.45μm, Prefilter, Double layers,100pcs/pk,10pk/cs.
Catalog No. NC2052765
Encompass_Preferred
Syringe Filter, PES, 13mm, 0.45μm, Prefilter, Double layers,100pcs/pk,10pk/cs.
Catalog No. NC2052765 Supplier WELCH MATERIALS INC Supplier No. S13PES045B10
Syringe Filter, PES, 13mm, 0.45μm, Prefilter, Double layers,100pcs/pk,10pk/cs.