Learn More
WELCH MATERIALS INC Welchrom Syringe Filter, Hydrophilic PTFE, 13mm, 0.22μm, Prefilter, Double layers, 100pcs/pk, 10pk/cs.
Catalog No. NC2052780
Encompass_Preferred
Syringe Filter, Hydrophilic PTFE, 13mm, 0.22μm, Prefilter, Double layers, 100pcs/pk, 10pk/cs.
Catalog No. NC2052780 Supplier WELCH MATERIALS INC Supplier No. S13PTL022B10
Syringe Filter, Hydrophilic PTFE, 13mm, 0.22μm, Prefilter, Double layers, 100pcs/pk, 10pk/cs.