Learn More
WELCH MATERIALS INC Welchrom Syringe Filter, Hydrophilic PVDF, 13mm, 0.22μm, Prefilter, Double layers, 100pcs/pk, 10pk/cs.
Supplier:  WELCH MATERIALS INC S13PVL022B10
Encompass_Preferred
Syringe Filter, Hydrophilic PVDF, 13mm, 0.22μm, Prefilter, Double layers, 100pcs/pk, 10pk/cs.
Catalog No. NC2052790