Learn More
WELCH MATERIALS INC Welchrom Syringe Filter, Hydrophilic PVDF, 25mm, 0.45μm, Prefilter, Double layers, 100pcs/pk, 10pk/cs.
Catalog No. NC2052795
Encompass_Preferred
Syringe Filter, Hydrophilic PVDF, 25mm, 0.45μm, Prefilter, Double layers, 100pcs/pk, 10pk/cs.
Catalog No. NC2052795 Supplier WELCH MATERIALS INC Supplier No. S25PVL045B10
Syringe Filter, Hydrophilic PVDF, 25mm, 0.45μm, Prefilter, Double layers, 100pcs/pk, 10pk/cs.