Learn More
Med Vet International Pillcrusher Oral Medication Syringe, 60 mL, Catheter Tip Without Safety, 3305, 30/Case
Catalog No. 503043085
Encompass_Preferred
More complete and accurate dose administration No danger of residual cross-contamination Faster and easier than any other pill-crushing method Reduces clogged tubes because medications are crushed to a consistently fine powder Provides a safer and more efficient way to deliver dissolved medications
Catalog No. 50-304-3085 Supplier Med Vet International Supplier No. 446005CS
More complete and accurate dose administration No danger of residual cross-contamination Faster and easier than any other pill-crushing method Reduces clogged tubes because medications are crushed to a consistently fine powder Provides a safer and more efficient way to deliver dissolved medications