Learn More
Chemglass Life Sciences Syringe, Polypropylene, Luer Slip, 10mL, Centric Tip, Non-Sterile, Bulk
SDP
Catalog No. 501531578
Encompass_Preferred
Syringe, Polypropylene, Luer Slip, 10mL, Offset Tip, Non-Sterile, Bulk. Dispenser packs of non-sterile syringes make sense. They're more economical and easier on the environment. These all polypropylene syringes are latex-free and contain no rubber or silicone. The plastic plunger has a smooth draw and a positive safety stop to prevent accidental spills. The 10mL sizes have a luer tip which is offset from the barrel. Pictured are the luer slip and luer lock styles, for reference.
Catalog No. 50-153-1578 Supplier Chemglass Life Sciences Supplier No. CG308204
 Restricted Item
Add to Cart
Add to Cart
Syringe, Polypropylene, Luer Slip, 10mL, Offset Tip, Non-Sterile, Bulk. Dispenser packs of non-sterile syringes make sense. They're more economical and easier on the environment. These all polypropylene syringes are latex-free and contain no rubber or silicone. The plastic plunger has a smooth draw and a positive safety stop to prevent accidental spills. The 10mL sizes have a luer tip which is offset from the barrel. Pictured are the luer slip and luer lock styles, for reference.