Learn More
Chemglass Life Sciences Syringe, Polypropylene, Luer Slip, 3mL, Centric Tip, Non-Sterile, Bulk
SDP
Catalog No. 501531576
Encompass_Preferred
Syringe, Polypropylene, Luer Slip, 3mL, Centric Tip, Non-Sterile, Bulk. Dispenser packs of non-sterile syringes make sense. They're more economical and easier on the environment. These all polypropylene syringes are latex-free and contain no rubber or silicone. The plastic plunger has a smooth draw and a positive safety stop to prevent accidental spills. The luer tip for the 3mL sizes is centric to the barrel. Pictured are the luer slip and luer lock styles, for reference.
Catalog No. 50-153-1576 Supplier Chemglass Life Sciences Supplier No. CG308202
Syringe, Polypropylene, Luer Slip, 3mL, Centric Tip, Non-Sterile, Bulk. Dispenser packs of non-sterile syringes make sense. They're more economical and easier on the environment. These all polypropylene syringes are latex-free and contain no rubber or silicone. The plastic plunger has a smooth draw and a positive safety stop to prevent accidental spills. The luer tip for the 3mL sizes is centric to the barrel. Pictured are the luer slip and luer lock styles, for reference.