Learn More
Chemglass Life Sciences Syringe, Polypropylene, 1mL Capacity, Centric Tip
SDP
Catalog No. CG308001
Encompass_Preferred
Syringe, Polypropylene, 1mL Capacity, Centric Tip. These all-polypropylene syringes are latex free and contain no rubber or silicone. The plastic plunger has a smooth draw and a positive safety stop to prevent accidental spills. Syringes are individually sterile strip packed. The Luer tip for the 1mL sizes is centric to the barrel. The 1mL syringe is a long, thin Tuberculin style.
Catalog No. CG308001 Supplier Chemglass Life Sciences Supplier No. CG308001
Syringe, Polypropylene, 1mL Capacity, Centric Tip. These all-polypropylene syringes are latex free and contain no rubber or silicone. The plastic plunger has a smooth draw and a positive safety stop to prevent accidental spills. Syringes are individually sterile strip packed. The Luer tip for the 1mL sizes is centric to the barrel. The 1mL syringe is a long, thin Tuberculin style.