Learn More
Chemglass Life Sciences Syringe, Polypropylene, 3mL Capacity, Centric Tip
SDP
Catalog No. CG308002
Encompass_Preferred
Syringe, Polypropylene, 3mL Capacity, Centric Tip. These all-polypropylene syringes are latex free and contain no rubber or silicone. The plastic plunger has a smooth draw and a positive safety stop to prevent accidental spills. Syringes are individually sterile strip packed. The Luer tip for the 3mL sizes is centric to the barrel.
Catalog No. CG308002 Supplier Chemglass Life Sciences Supplier No. CG308002
 Restricted Item
Add to Cart
Add to Cart
Syringe, Polypropylene, 3mL Capacity, Centric Tip. These all-polypropylene syringes are latex free and contain no rubber or silicone. The plastic plunger has a smooth draw and a positive safety stop to prevent accidental spills. Syringes are individually sterile strip packed. The Luer tip for the 3mL sizes is centric to the barrel.