Learn More
Chemglass Life Sciences Syringe, Polypropylene, 10mL Capacity, Offset Tip
SDP
Catalog No. CG308006
Encompass_Preferred
Syringe, Polypropylene, 10mL Capacity, Offset Tip. These all-polypropylene syringes are latex free and contain no rubber or silicone. The plastic plunger has a smooth draw and a positive safety stop to prevent accidental spills. Syringes are individually sterile strip packed. The Luer tip for 10mL sizes have a Luer tip which is offset from the barrel.
Catalog No. CG308006 Supplier Chemglass Life Sciences Supplier No. CG308006
Syringe, Polypropylene, 10mL Capacity, Offset Tip. These all-polypropylene syringes are latex free and contain no rubber or silicone. The plastic plunger has a smooth draw and a positive safety stop to prevent accidental spills. Syringes are individually sterile strip packed. The Luer tip for 10mL sizes have a Luer tip which is offset from the barrel.