Learn More
Chemglass Life Sciences Syringe, Polypropylene, 20mL Capacity, Offset Tip
SDP
Catalog No. CG308008
Encompass_Preferred
Syringe, Polypropylene, 20mL Capacity, Offset Tip. These all-polypropylene syringes are latex free and contain no rubber or silicone. The plastic plunger has a smooth draw and a positive safety stop to prevent accidental spills. Syringes are individually sterile strip packed. The Luer tip for the 20mL sizes have a Luer tip which is offset from the barrel.
Catalog No. CG308008 Supplier Chemglass Life Sciences Supplier No. CG308008
 Restricted Item
Add to Cart
Add to Cart
Syringe, Polypropylene, 20mL Capacity, Offset Tip. These all-polypropylene syringes are latex free and contain no rubber or silicone. The plastic plunger has a smooth draw and a positive safety stop to prevent accidental spills. Syringes are individually sterile strip packed. The Luer tip for the 20mL sizes have a Luer tip which is offset from the barrel.