Learn More
Sigma Aldrich Fine Chemicals Biosciences Syringe PP/PE without need
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z683566100EA
Encompass
These polypropylene syringes with Luer tips may be used with any size needle. The rigid polyethylene plungers of these disposable syringes are slightly oversized creating a tight seal against the walls of the semi-flexible PP barrel as well as providing an exceptionally smooth draw. No contaminating lubricants added.Syringes are individually packaged (sterile) except for bulk packed products these are supplied in plastic bags (non-sterile).Order needles separately.
Catalog No. 50-290-1524