Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Touch-Screen Syringe Pump, Infusion, One-Syringe, 1.28 pL/min to 88.28 mL/min; 100 to 240 VAC
Catalog No. NC1564958
Encompass
These entry-level pumps offer features normally found in more expensive models. An intuitive touch-screen display lets you see all operating parameters and easily create and recall configurations. Front-panel LED light indicates when pump is operating¿vie
Catalog No. NC1564958 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7490502
These entry-level pumps offer features normally found in more expensive models. An intuitive touch-screen display lets you see all operating parameters and easily create and recall configurations. Front-panel LED light indicates when pump is operating¿vie